PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 36×36×10 mm
  77908-056
New Product
 :   StatLab
 :  GBC-3614-05A
PathFlow Supa Mega Base Mold
  77908-056
 :  StatLab
New Product
PathFlow Supa Mega Base Mold
PathFlow Supa Mega base mold, 36×36×10 mm
$242.70
Pack of 5
PathFlow stainless steel Supa Mega base molds.
  • Reusable Stops wax leakage Stackable design for efficient storage Stainless steel for a lifetime of use
  • Désignation:
    PathFlow Supa Mega base mold
  • Dimensions:
    36×36×10 mm