Clearview Classroom Stream Table
Product Details & Documents
Simulate a flowing stream and study erosion with this classroom stream table.
Clear surface for visual study
Ideal for environmental and water studies
Water tight
Easy to store
Bring Water Dynamics to Life with the Clearview Classroom Stream Table - a must have for any earth science classroom! This interactive stream table offers a dynamic way for students to explore erosion, sediment transport, and river formation. Designed for easy setup and durability, the rugged ABS construction features an all-in-one system—just add water, activate the pump, and start experimenting. Five adjustable water jets simulate natural water flow, from small streams to converging trib. Ships oversize.
Spécifications
- Désignation:Replacement pump