Clearview Classroom Stream Table
Clearview Classroom Stream Table
Replacement pump
Catalog # 470358-450
New Product
Supplier:   Science First
Supplier #:  035590
Clearview Classroom Stream Table
Catalog # 470358-450
Supplier:  Science First
New Product
Clearview Classroom Stream Table
Clearview Classroom Stream Table
Replacement pump
Price
$63.12
Log in pour la disponibilité
Each
Product Details & Documents
Simulate a flowing stream and study erosion with this classroom stream table.
  • Clear surface for visual study
    Ideal for environmental and water studies
    Water tight
    Easy to store
Bring Water Dynamics to Life with the Clearview Classroom Stream Table - a must have for any earth science classroom! This interactive stream table offers a dynamic way for students to explore erosion, sediment transport, and river formation. Designed for easy setup and durability, the rugged ABS construction features an all-in-one system—just add water, activate the pump, and start experimenting. Five adjustable water jets simulate natural water flow, from small streams to converging trib. Ships oversize.
Spécifications
  • Désignation:
    Replacement pump