2ACTAMD-2-DEOXY-346-TRI-O 250MG
Catalog # 77888-976
Supplier: AFG Bioscience
New Product
2ACTAMD-2-DEOXY-346-TRI-O 250MG
Catalog # 77888-976
Supplier: AFG Bioscience
Supplier Number: C54141-250MG
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
MICROLITER® 700 Series Syringes, Hamilton
Supplier: Hamilton
Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
Single-Use Needles, BD Medical
Supplier: BD
Toluène ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Acétonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
TelN Protelomerase
Supplier: New England Biolabs (NEB)
Q qPCR Instrument, Quantabio
Supplier: Quantabio
Ether diéthylique ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Potassium chloride 2 M in aqueous solution, sterile
Supplier: Teknova