N N N N-TETRAACETYLCHITOTETRAOSE 5MG
Catalogue # 77888-638
Fournisseur: AFG Bioscience
New Product
N N N N-TETRAACETYLCHITOTETRAOSE 5MG
Catalogue # 77888-638
Fournisseur: AFG Bioscience
Supplier Number: C50356-5MG
New Product
Frequently Bought Together
American® White Cross Adhesive Bandages, DUKAL™ Corporation
Fournisseur: Dukal
Optical Lens Cleaner, FoxWest Sales
Fournisseur: Decon Labs
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Fournisseur: VWR International
Vesphene® III st Disinfectant Cleaners
Fournisseur: Steris
Potassium dihydrogen phosphate ≥99% (by acidimetry), GR ACS, Supelco®
Fournisseur: MilliporeSigma
Sodium chlorure ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Fournisseur: Air-Tite
VWR® Traceable® Conductivity Calibration Standards (CRM)
Fournisseur: VWR International
Liquinox® Critical Cleaning Liquid Detergents
Fournisseur: Alconox
pH Test Strips, VWR Chemicals BDH®
Fournisseur: BDH
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Leakbuster™ Specimen Containers
Fournisseur: Starplex
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
Components and Accessories for Miele Lab Washers, Miele Professional
Fournisseur: Miele
Trimming Blades, Avantik
Fournisseur: Avantik
ACETIC RINSE SOLUTION 4L
Fournisseur: Ostrem Chemical
SporAmpule® Liquid Biological Indicator
Fournisseur: HARDY DIAGNOSTICS CA
Zinc Chloride
Fournisseur: Ward's Science