CytoFLEX Mosaic Detection Modules | Avantor
CytoFLEX Mosaic Detection Modules
CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)
Catalog # CA77886-988
Supplier:   Beckman Coulter
Fournisseur #:  D20165
CytoFLEX Mosaic Detection Modules
Catalog # CA77886-988
Supplier:  Beckman Coulter
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Spécifications
  • Désignation:
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)