Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Human Recombinant IL-18 (from E. coli)

Vial, 0.1 mg
Catalog # 77870-860
Supplier:   Sartorius
 :  CYK-0100-1015
Human Recombinant IL-18 (from E. coli)
Catalog # 77870-860
Supplier:  Sartorius
CAS Number:  
Human Recombinant IL-18 (from E. coli)
Human Recombinant IL-18 (from E. coli)

Vial, 0.1 mg

CAS Number:  
Price
$808.91
Log in pour la disponibilité
Each
 
Product Details & Documents

Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Taille:
    0.1 mg
  • Protease-free:
    N
  • Protein/peptide name:
    IL-18
  • Protein synonyms:
    Interleukin-18
  • Protein Function:
    Cytokine
  • Source:
    E. coli
  • Shipping temperature:
    Ambient
  • Nuclease-free:
    N
  • Cat. No.:
    77870-860
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Ambient
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    Y
  • Species:
    Human
  • Animal-free:
    Y

Spécifications