Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant IL-18 (from E. coli) | Avantor

Human Recombinant IL-18 (from E. coli)

Vial, 0.025 mg
Catalog # 77870-807
Supplier:   Sartorius
Fournisseur #:  CYK-0025-1015
Human Recombinant IL-18 (from E. coli)
Catalog # 77870-807
Supplier:  Sartorius
CAS Number:  
Human Recombinant IL-18 (from E. coli)
Human Recombinant IL-18 (from E. coli)

Vial, 0.025 mg

CAS Number:  
Price
$309.52
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Taille:
    0.025 mg
  • Protease-free:
    N
  • Protein/peptide name:
    IL-18
  • Protein synonyms:
    Interleukin-18
  • Protein Function:
    Cytokine
  • Source:
    E. coli
  • Shipping temperature:
    Ambient
  • Nuclease-free:
    N
  • Cat. No.:
    77870-807
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Ambient
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    Y
  • Species:
    Human
  • Animal-free:
    Y

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.