Zero Waste Box™ Plastic Packaging Recycling Boxes
Zero Waste Box™ Plastic Packaging Recycling Boxes
Plastic packaging Zero Waste Box™, Cardboard, Large, 15×15×37"
Catalog # 7786767-918
Supplier:   Terracycle
Supplier #:  CS33-L
Zero Waste Box™ Plastic Packaging Recycling Boxes
Catalog # 7786767-918
Supplier:  Terracycle
Zero Waste Box™ Plastic Packaging Recycling Boxes
Zero Waste Box™ Plastic Packaging Recycling Boxes
Plastic packaging Zero Waste Box™, Cardboard, Large, 15×15×37"
Price
$404.96
Log in for availability
Each
Product Details & Documents
This easy-to-use recycling program provides a complete waste collection, shipment, and recycling solution for any flexible or rigid packaging.
  • Reduce landfill waste
  • Support your company's sustainability goals
  • Each collection box can be placed at convenient locations throughout the lab or facility
  • When full, simply place the collection box in a designated shipping area
  • Each box has a pre-paid UPS shipping label affixed
This program offers a convenient opportunity to recycle various types of waste found in most labs. Do not send in shipping and packaging peanuts, biodegradable PLA plastics, plastic with paper stickers, and please remove food crumbs, food waste, and plastic with heavy contamination prior to sending. Any item contaminated with hazardous, flammable, or infectious substances are not accepted in this box.
Each sturdy collection box is made of double-corrugated cardboard. Boxes are supplied with simple assembly instructions and a pre-affixed return shipping label.
Order: Select a Zero Waste Box™ and place an order.
Collect: Collect the accepted items to fill the box.
Ship: Once full, place the box at an outbound UPS location for shipping using the pre-paid shipping label (already fastened to the box).
Reorder: Order the next Zero Waste Box™ to continue to recycle everything.
Certifications: SFI (license code: SGSNA-SFICOC-603839); FSC (license code: SGSCH-COC-000508); Bureau Veritas.
Specifications
  • Désignation:
    Plastic packaging Zero Waste Box™
  • Dimensions:
    15×15×37"
  • Matière:
    Cardboard
  • Taille:
    Large
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.