Avantor® PUPSIT Assemblies | Avantor
Avantor® PUPSIT Assemblies
PUPSIT assembly with Cytiva Supor filters
Catalog # 77865-530
Fournisseur #:  1014904
Avantor® PUPSIT Assemblies
Catalog # 77865-530
Avantor® PUPSIT Assemblies
Avantor® PUPSIT Assemblies
PUPSIT assembly with Cytiva Supor filters
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Pack of 5
Product Details & Documents
Avantor's PUPSIT (Pre-Use Post-Sterilization Integrity Testing) assemblies are designed to simplify pre-use integrity testing while maintaining product sterility.
  • Configurable designs that easily integrate into your filtration and filling process
  • Reduced risks associated with designing and implementation of PUPSIT
  • Open architecture for greater flexibility in configuration and enhanced supply chain security
    Faster lead times compared to fully customized assembly solutions
We offer a range of pre-designed PUPSIT assemblies with configurable elements. By leveraging our extensive library of validated components and a supplier-agnostic approach, we enable flexible design adaptation while maintaining compliance with EU GMP Annex 1. Choose from a large validated component library that includes:
• Filters - air filter, liquid filter, barrier filter
• Connectors - quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Tubing - thermoplastic elastomers, platinum-cured silicone, reinforced tubing
• Sensors - sensors or single-use guage tees
• Fittings - straight, reducing, elbow, T-style, X-style, and cross-style
• Sample ports - septum-style, EVSS, Omnitop, other needle-based and needle-free ports
Certifications: PUPSIT assemblies are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. All product contact materials are animal-free or processed in accordance to EMEA/410/01. These assemblies are gamma irradiated to Sterility Assurance Level (SAL) 10-6. Each assembly is individually triple bagged and is accompanied with a certificate of quality and certificate of processing.
Ordering information: PUPSIT assemblies are available in a wide variety of configurable designs, as well as custom engineered designs, to suit specific applications.   Few options are presented below. For more information, please contact a local Avantor sales representative.
Spécifications
  • Désignation:
    PUPSIT assembly with Cytiva Supor filters
  • Filtration area:
    0.6 m²
  • Cat. No.:
    77865-530
  • Tubing:
    Reinforced silicone
  • Filter:
    Liquid filter: Cytiva Supor / Barrier filter: Zebragard Barrier
  • Sensor:
    Single-use Gauge Tee (SGT)