Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dog Recombinant GM-CSF (from E. coli)

Dog Recombinant GM-CSF (from E. coli)

Vial, 100 µg
Catalog # 77753-268
Supplier:   VWR International
Dog Recombinant GM-CSF (from E. coli)
Catalog # 77753-268
Supplier:  VWR International
CAS Number:  
Dog Recombinant GM-CSF (from E. coli)
Dog Recombinant GM-CSF (from E. coli)

Vial, 100 µg

CAS Number:  
Price
$1,232.40
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications

Specifications

  • Type de conditionnement:
    Vial
  • Taille:
    100 µg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    14.4 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Gene ID:
    P48749
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Dog
  • Protein/peptide name:
    GM-CSF
  • Protein synonyms:
    CSF-2|Pluripoietin-α|MGI-1GM
  • Protein Function:
    Cytokine
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MAPTRSPTLV TRPSQHVDAI QEALSLLNNS NDVTAVMNKA VKVVSEVFDP EGPTCLETRL QLYKEGLQGS LTSLKNPLTM MANHYKQHCP PTPESPCATQ NINFKSFKEN LKDFLFNIPF DCWKPVKK
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤15 ng/ml; Bioactivity ≥6.7×10⁴ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    77753-268
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    −20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    Y

Specifications