Mouse Recombinant RELM-A (from E. coli)
Vial, 5 µg
77752-218
Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Type de conditionnement:Vial
- Taille:5 µg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:9.6 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Gene ID:Q9EP95
- Formulation:10 mM Sodium phosphate, pH 7.5
- Endotoxin-free:N
- Species:Mouse
- Protein/peptide name:RELM-A
- Protein synonyms:FIZZ1
- Protein Function:Cytokine
- Endotoxin level:Low
- Grade:RUO
- Sequence:MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
- Biological activity:Bioactivity: No biological activity data is available at this time
- Shipping temperature:RT
- Cat. No.:77752-218
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:−20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



