Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Recombinant RELM-A (from E. coli)

Mouse Recombinant RELM-A (from E. coli)

Vial, 500 µg
Catalog # 77752-216
Supplier:   VWR International
Mouse Recombinant RELM-A (from E. coli)
Catalog # 77752-216
Supplier:  VWR International
CAS Number:  
Mouse Recombinant RELM-A (from E. coli)
Mouse Recombinant RELM-A (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$2,788.70
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications

Specifications

  • Type de conditionnement:
    Vial
  • Taille:
    500 µg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    9.6 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Gene ID:
    Q9EP95
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Endotoxin-free:
    N
  • Species:
    Mouse
  • Protein/peptide name:
    RELM-A
  • Protein synonyms:
    FIZZ1
  • Protein Function:
    Cytokine
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Shipping temperature:
    RT
  • Cat. No.:
    77752-216
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    −20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications