Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Recombinant PDGF-BB (from E. coli cells) | Avantor

Mouse Recombinant PDGF-BB (from E. coli cells)

Vial, 100 µg
Catalog # 77752-480
Supplier:   VWR International
Mouse Recombinant PDGF-BB (from E. coli cells)
Catalog # 77752-480
Supplier:  VWR International
CAS Number:  
Mouse Recombinant PDGF-BB (from E. coli cells)
Mouse Recombinant PDGF-BB (from E. coli cells)

Vial, 100 µg

CAS Number:  
Price
$732.06
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β, and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.

  • High quality
  • Low endotoxin
  • Affordable

PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.

Caution: For research use only.

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Taille:
    100 µg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    12.4/24.7 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Gene ID:
    P31240
  • Formulation:
    10 mM sodium citrate, pH 3.0
  • Endotoxin-free:
    N
  • Species:
    Mouse
  • Protein/peptide name:
    PDGF-BB
  • Protein synonyms:
    GDGF|ODGF
  • Protein Function:
    Cytokine
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MSLGSLAAAE PAVIAECKTR TEVFQISRNL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRASQVQMRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETIVTPRPVT
  • Biological activity:
    Bioactivity assay: 3T3 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
  • Shipping temperature:
    Room temperature
  • Cat. No.:
    77752-480
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.