Mouse Recombinant SCF (from E. coli)
Vial, 10 µg
Catalog # 77751-988
Supplier: VWR International
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Spécifications
Spécifications
- Type de conditionnement:Vial
- Taille:10 µg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:18.4 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Gene ID:P20826
- Formulation:20 mM Sodium phosphate, pH 6.5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Mouse
- Protein/peptide name:SCF (AF)
- Protein synonyms:c-Kit Ligand|KL|Steel Factor|MGF
- Protein Function:Cytokine
- Endotoxin level:Low
- Grade:RUO
- Sequence:MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
- Shipping temperature:RT
- Cat. No.:77751-988
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:Y



