Human Recombinant CNTF (from E. coli)
Vial, 5 µg
77751-646
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Type de conditionnement:Vial
- Taille:5 µg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:22.9 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Gene ID:P26441
- Formulation:10 mM Sodium phosphate, pH 7.5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Human
- Protein/peptide name:CNTF (AF)
- Protein Function:Cytokine
- Endotoxin level:Low
- Grade:RUO
- Sequence:MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤325 ng/ml; Bioactivity ≥3.1×10³ units/mg
- Shipping temperature:RT
- Cat. No.:77751-646
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:Y
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



