Human Recombinant CNTF (from E. coli)
Vial, 20 µg
77751-642
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Type de conditionnement:Vial
- Taille:20 µg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:22.9 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Gene ID:P26441
- Formulation:10 mM Sodium phosphate, pH 7.5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Human
- Protein/peptide name:CNTF (AF)
- Protein Function:Cytokine
- Endotoxin level:Low
- Grade:RUO
- Sequence:MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤325 ng/ml; Bioactivity ≥3.1×10³ units/mg
- Shipping temperature:RT
- Cat. No.:77751-642
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:Y



