Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant CNTF (from E. coli) | Avantor

Human Recombinant CNTF (from E. coli)

Vial, 100 µg
Catalog # 77751-638
Supplier:   VWR International
Human Recombinant CNTF (from E. coli)
Catalog # 77751-638
Supplier:  VWR International
CAS Number:  
Human Recombinant CNTF (from E. coli)
Human Recombinant CNTF (from E. coli)

Vial, 100 µg

CAS Number:  
Price
$716.26
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Taille:
    100 µg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    22.9 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Gene ID:
    P26441
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Human
  • Protein/peptide name:
    CNTF (AF)
  • Protein Function:
    Cytokine
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤325 ng/ml; Bioactivity ≥3.1×10³ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    77751-638
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    Y

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.