Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Recombinant RELM-A (from E. coli) | Avantor

Mouse Recombinant RELM-A (from E. coli)

Vial, 1 mg
Catalog # 77752-212
Supplier:   VWR International
Mouse Recombinant RELM-A (from E. coli)
Catalog # 77752-212
Supplier:  VWR International
CAS Number:  
Mouse Recombinant RELM-A (from E. coli)
Mouse Recombinant RELM-A (from E. coli)

Vial, 1 mg

CAS Number:  
Price
$4,645.20
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.

  • High quality

  • Low endotoxin
  • Affordable

Caution: For research use only.

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Taille:
    1 mg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    9.6 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Gene ID:
    Q9EP95
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Endotoxin-free:
    N
  • Species:
    Mouse
  • Protein/peptide name:
    RELM-A
  • Protein synonyms:
    FIZZ1
  • Protein Function:
    Cytokine
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Shipping temperature:
    RT
  • Cat. No.:
    77752-212
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    −20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Spécifications