Human Recombinant GM-CSF (from E. coli cells)
Vial, 1 mg
Catalog # 77750-110
Supplier: VWR International
Human Recombinant GM-CSF (from E. coli cells)
Catalog # 77750-110
Supplier: VWR International
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells.
- High quality
- Low endotoxin
- Affordable
GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
Caution: For research use only.
Spécifications
Spécifications
- Type de conditionnement:Vial
- Taille:1 mg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:14.6 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Gene ID:P04141
- Formulation:10 mM sodium phosphate, pH 7.5
- Endotoxin-free:N
- Species:Human
- Protein/peptide name:GM-CSF
- Protein synonyms:CSF-2|Pluripoietin-α|MGI-1GM
- Protein Function:Cytokine
- Endotoxin level:Low
- Grade:RUO
- Sequence:MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤0.2 ng/ml; Bioactivity ≥5.0×10⁶ units/mg
- Shipping temperature:RT
- Cat. No.:77750-110
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



