Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sf9 Cells

Sf9 Cells

Sf9 cells, suspension in ESF AF, approx. 1 M cells/ml (50/125 ml flask), Proliferative
Catalog # 77705-762
Supplier #:  94-006S
Sf9 Cells
Catalog # 77705-762
CAS Number:  
Sf9 Cells

Sf9 cells, suspension in ESF AF, approx. 1 M cells/ml (50/125 ml flask), Proliferative

CAS Number:  
Price
$386.83
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.

  • Recombinant protein expression from a variety of expression platforms
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Quality and performance testing

The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.

The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.

Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.

Delivery information: Priority overnight.

Specifications

Specifications

  • Format de cellule:
    Proliferative
  • Désignation:
    Sf9 cells, suspension in ESF AF, approx. 1 M cells/ml (50/125 ml flask)

Specifications