Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sf9 Cells

Sf9 Cells

Sf9 cells, frozen in ESF 921, 50 M cells/vial, Cryopreserved
Catalog # 77705-746
Supplier #:  94-001F
Sf9 Cells
Catalog # 77705-746
CAS Number:  
Sf9 Cells

Sf9 cells, frozen in ESF 921, 50 M cells/vial, Cryopreserved

CAS Number:  
Price
$857.66
Log in pour la disponibilité
Each
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.

  • Recombinant protein expression from a variety of expression platforms
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Quality and performance testing

The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.

The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.

Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.

Delivery information: Priority overnight.

Spécifications

Spécifications

  • Format de cellule:
    Cryopreserved
  • Désignation:
    Sf9 cells, frozen in ESF 921, 50 M cells/vial

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.