Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Buffer Solutions

Buffer Solutions

Buffer solution, 1.68
Catalog # 77679-404
Supplier:   Ohaus
Supplier #:  30100424
Buffer Solutions
Catalog # 77679-404
Supplier:  Ohaus
CAS Number:  
Buffer Solutions
Buffer Solutions

Buffer solution, 1.68

CAS Number:  
Price
$53.57
Log in for availability
Each
Product Details & Documents

Buffer solution, 250 ml bottle.

Specifications

Specifications

  • pH:
    1.68
  • Density:
    Buffer solution

Specifications