Buffer Solutions
Buffer solution, 4.01
77679-406
: Ohaus
: 30100425
Buffer solution, 250 ml bottle.
- pH:4.01
- Density:Buffer solution
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



