Buffer Solutions
Buffer solution, 7.00
Catalog # 77679-410
Supplier: Ohaus
Fournisseur #: 30100427
Jump to:
Product Details & Documents
Buffer solution, 250 ml bottle.
Spécifications
Spécifications
- pH:7.00
- Density:Buffer solution
- Cat. No.:77679-410



