Ohaus® Starter Electrodes
Product Details & Documents
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes.
- Can be used in conjunction with Ohaus® Starter Series
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes that can be used in conjunction with Ohaus® Starter Series 2100 and 3100 bench and Ohaus® Starter 300 Series Portable meters.
Ordering information: Ohaus® 2100 & 3100 Starter Bench Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 3100C Starter Bench Meter Compatible Electrodes are STCON3. Ohaus® 300 Starter Portable Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 300C Starter Portable Meter Compatible Electrodes are STCON3. Ohaus® 300D Starter Portable Meter Compatible Electrodes are STTEMP30, STD011.
Spécifications
- Désignation:ST310: General purpose 3-in-1 plastic refillable pH electrode
- For Use With:Series 300, 2100, 3100
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Maintenance kit