SAMPLING MANIFOLD 5 X 15ML PP
Catalog # 77675-076
Supplier: AVANTOR FLUID HANDLING, LLC
New Product
SAMPLING MANIFOLD 5 X 15ML PP
Catalog # 77675-076
Supplier: AVANTOR FLUID HANDLING, LLC
Supplier Number: 1013605
New Product
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
VWR® Lab Wrap, Clear Plastic
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Microscope Immersion Oil, Cargille
Supplier: Cargille Laboratories
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
Ethyl Alcohol Denatured
Supplier: Ward's Science
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
Theobromine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ S-Nitrosylation Western Blot Kit, Thermo Scientific
Supplier: Thermo Scientific
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach
Formalin 10%, neutral buffered, VWR®
Supplier: VWR International
Acide chlorhydrique 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC