Sterilisation Baskets, Side Perforated, Flat Base | Avantor
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 120×250×50 mm
Catalog # 77666-140
Fournisseur #:  WP-4545
Sterilisation Baskets, Side Perforated, Flat Base
Catalog # 77666-140
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 120×250×50 mm
Price
$97.96
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Spécifications
  • Dimensions:
    120×250×50 mm
  • Matière:
    Stainless steel 304