Vitrectomy Scissors | Avantor
Vitrectomy Scissors
Vitrectomy scissors, 65°, 20 G, Stainless steel, Angled
Catalog # 77666-308
Fournisseur #:  WP504240
Vitrectomy Scissors
Catalog # 77666-308
Vitrectomy Scissors
Vitrectomy scissors, 65°, 20 G, Stainless steel, Angled
Price
778,94 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vitrectomy scissors are pencil-style scissors with a new, ergonomic handle design with micro-blades that are easy to clean. Two models available, straight and angled. These scissors designed for vitrectomy, an ophthalmalogical procedure.
Vitrectomy is the surgical removal of some or all of the vitreous humor, the gel that fills the eyeball, in order to correct retinal issues like retinal tears, macular holes, floaters and more. During surgery, an ophthalmologist penetrates the white of the eye, the sclera, and inserts the instruments necessary to complete the surgery.
Spécifications
  • Désignation:
    Vitrectomy scissors, 65°, 20 G
  • Matière:
    Stainless steel
  • Blade Type:
    Angled