Sterilization Baskets, Side Perforated, Woven Wire Base
Product Details & Documents
The sterilization basket is an ideal solution for instrument cleaning, sterilization and storage.
- Manufactured with stainless steel 304, non-magnetic
- Electropolished
- Burr free to protect wraps and prevent injury
- Smooth rounded corners
- Single frame, double frame and detention frame lids available
Side walls are perforated to protect sharp instrument edges and tips from damage. The hole size is 2.0 mm. Meets all the requirements of automatic washing and sterilization for all types of instruments. Separate lids are available with single frame, double frame and detention shapes. Available with drop in and folding handles.
Top and bottom frame diameter: 5.0 mm. Base available in wire mesh with opening squares of 4.2 mm and wire diameter of 1.0 mm.
Spécifications
- Désignation:Sterilization baskets
- Dimensions:540×250×70 mm
- Matière:Stainless steel 304