Sterilisation Baskets, Side Perforated, Flat Base
Product Details & Documents
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Spécifications
- Dimensions:480×250×70 mm
- Matière:Stainless steel 304