Microelectrode Holders, MPH6S | Avantor
Microelectrode Holders, MPH6S
2.0 mm standard single capillary tubing, Microelectrode holder (MPH6S)
Catalog # 77664-880
Fournisseur #:  MPH6S20
Microelectrode Holders, MPH6S
Catalog # 77664-880
Microelectrode Holders, MPH6S
2.0 mm standard single capillary tubing, Microelectrode holder (MPH6S)
Price
$143.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Female luer
  • Screw cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. The holder style you select will depend on your experimental application, space, and instrumentation.
Spécifications
  • Désignation:
    Microelectrode holder (MPH6S)
  • For Use With:
    2.0 mm standard single capillary tubing