Microelectrode Holders | Avantor
Microelectrode Holders
Microelectrode holder, size: 2.0 mm
Catalog # 77664-860
Fournisseur #:  MPH420
Microelectrode Holders
Catalog # 77664-860
Microelectrode Holders
Microelectrode holder, size: 2.0 mm
Price
$143.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Male connector
  • Pellet half-cell
  • No pressure port
  • Screw cap
Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. The holder style selected will depend on the experimental application, space, and instrumentation.
Delivery information: All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Spécifications
  • Désignation:
    Microelectrode holder, size: 2.0 mm