Microelectrode Holders
1.2 mm standard WPI single capillary tubing
  77664-640
 :  MEH2RW12
Microelectrode Holders
  77664-640
Microelectrode Holders
1.2 mm standard WPI single capillary tubing
$200.68
Each
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Male connector
  • Wire half-cell
  • Male luer
  • Screw cap
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • For Use With:
    1.2 mm standard WPI single capillary tubing