Dri-Ref™ Reference Electrodes
KCl
Catalog # 77664-220
 :  DRIREF-450
Dri-Ref™ Reference Electrodes
Catalog # 77664-220
Dri-Ref™ Reference Electrodes
KCl
Price
$840.56
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dri-Ref™ reference electrodes have extremely low electrolyte leak­age properties and exhibit stable and reproducible potential and low resistance.
  • Stored in KCl when not in use, they have a long life expectancy
  • Extremely low electrolyte leakage
  • Stable, reproducible potential with low resistance
  • Chemically resistant to strong acids and bases
  • Ideal for small volume, low salt concentration measurement (SUPER-Dri-Ref)
May be used with ion selective electrodes without contamination from the reference electrode. Although the internal filling solution contains KCI, the low fluid leakage means Dri-Ref may be used in combination with ion selective electrodes, including those for K+ and Cl-, without significant contamination from the reference electrode.
Dri-Ref electrodes are not suitable for use in organic solvents. In addition, the long, thin FLEXREF may be easily manipulated to accommodate a difficult experimental setup.
SUPER-Dri-Ref - With a diameter of 2 mm, SUPER-Dri-Ref does not leak electrolyte at all. Exhibiting the electrical stability of a classic flow­ing junction reference cell, this electrode exhibits low re­sis­tance and a stable half-cell po­ten­tial es­sen­tial­ly in­de­pen­dent of sample electrolyte concentration. SUPER-Dri-Ref is ideal for small vol­ume and low salt concentration mea­sure­ments.
DRIREF-450 - Micro-reference electrode – Only 450 µm in diameter and one inch long, this reference electrode can be used along with other sensors in space-restricted areas and very small sample volumes.
DRIREF-L - The male Luer fitting at the front of the electrode allows it to be easily connected to a female Luer port to form a tight seal - a very convenient installation for a flow-through system.
Spécifications
  • Fill Solution:
    KCl