Reference Cell Electrodes | Avantor
Reference Cell Electrodes
Reference cell with 4' lead
Catalog # 77665-100
Fournisseur #:  RC1
Reference Cell Electrodes
Catalog # 77665-100
Reference Cell Electrodes
Reference cell with 4' lead
Price
$180.12
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
  • Reference cell electrodes for laboratory research
Spécifications
  • Désignation:
    Reference cell with 4' lead