Microelectrode Holders
Microelectrode Holders
1.0 mm standard WPI single capillary tubing
  77664-606
Supplier #:  MEH1R10
Microelectrode Holders
  77664-606
Microelectrode Holders
1.0 mm standard WPI single capillary tubing
$142.20
Each
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Male connector
  • Pellet half-cell
  • No pressure port, no cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • For Use With:
    1.0 mm standard WPI single capillary tubing