Wire Mesh Baskets For Sterilization and Sorting
Product Details & Documents
In research laboratories, wire mesh baskets are commonly used in research laboratories, specifically for sterilization protocols, but they are practical for many other uses too.
- Smooth round edges
- Used for handling and storage of surgical instruments
Research laboratories frequently use wire mesh baskets to hold laboratory glassware, metal instruments, and other items during autoclaving and sterilization processes. The open design allows for efficient steam penetration, ensuring proper sterilization.
They help maintain a clean and organized workspace. After cleaning surgical instruments, wire mesh baskets are useful for air-drying. Use them to hold and sort laboratory samples. Their open design allows easy visual inspection, and they can be labeled for better organization.
Whether you are using them in an autoclave, your bench top or the freezer, wire mesh baskets in a variety of sizes are helpful around the lab. These are non-magnetic stainless steel, and they have smooth round edges.
Spécifications
- Dimensions:265×125×30 mm
- Matière:Non-magnetic stainless steel