Absorbent Paper Points | Avantor
Absorbent Paper Points
Blue, Medium, Pack of 200
Catalog # 77663-546
Fournisseur #:  504182
Absorbent Paper Points
Catalog # 77663-546
Absorbent Paper Points
Blue, Medium, Pack of 200
Price
$63.28
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Absorbent paper points are commonly used in laboratories, particularly in the field of dentistry and endodontics. These paper points are small, sterile, and highly absorbent.
  • Made from higly absorbent paper
  • Uniformly rolled to ensure correct size
  • Good for small volume absorption in confined areas
These sterile paper points are tapered to target specific areas and soak up small volumes of liquid. A variety of color-coded sizes are available, including extra fine, #15 fine, medium and coarse.
Packaging: Package of 100 to 200, depending on the size of the paper points.
Spécifications
  • Color:
    Blue
  • Packaging:
    Pack of 200
  • Type:
    Medium