Sterilization Mesh Cassettes | Avantor
Sterilization Mesh Cassettes
4×4×2 cm (1.5×1.5×.75")
Catalog # 77663-776
Fournisseur #:  504766
Sterilization Mesh Cassettes
Catalog # 77663-776
Sterilization Mesh Cassettes
4×4×2 cm (1.5×1.5×.75")
Price
$90.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished.
  • These baskets come with a wire locking clip
  • This style of sterilization baskets is available in three sizes.
They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm.
Spécifications
  • Dimensions:
    4×4×2 cm (1.5×1.5×.75")