Adson Forceps, Straight
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
: Package of 12.
- Désignation:Adson forcep, 1×2 teeth
- Longueur:12 cm (4.75")
- Matière:Stainless steel
- Curvature:Straight