Mini Iris Scissors | Avantor
Mini Iris Scissors
Mini iris scissor, sharp tips, 8 cm (3.1"), Stainless steel, Curved
Catalog # 77663-350
Fournisseur #:  503671
Mini Iris Scissors
Catalog # 77663-350
Mini Iris Scissors
Mini iris scissor, sharp tips, 8 cm (3.1"), Stainless steel, Curved
Price
$82.73
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Spécifications
  • Désignation:
    Mini iris scissor, sharp tips
  • Longueur:
    8 cm (3.1")
  • Matière:
    Stainless steel
  • Curvature:
    Curved