Micro Mosquito Forceps
Micro Mosquito Forceps
Stainless steel, Right angle
Catalog # 77663-282
Supplier #:  503360
Micro Mosquito Forceps
Catalog # 77663-282
Micro Mosquito Forceps
Stainless steel, Right angle
Price
$153.31
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro Mosquito Forceps, also known as microsurgical mosquito forceps or micro hemostatic forceps, are designed for extremely delicate and precise surgical procedures. Micro Mosquito Forceps are significantly smaller in size compared to regular mosquito forceps. These are 12.5 cm (5") long with fine tips. Looking at the end of the tip of one blade of these forceps, the dimensions are 0.4 mm thick and 0.8 mm wide.
  • 12.5 cm (5") Long
  • Serrated, fine (0.4×0.8 mm) tips
  • Straight, curved or right angle tips
  • Surgical grade stainless steel or premium German steel
You can count on our Micro Mosquito Forceps for precise manipulation of very small tissues or delicate structures in tight spaces. The tips of Micro Mosquito Forceps are extremely thin, enabling them to grasp and manipulate tiny structures with precision. Available tips include straight, curved, and right angled. Micro Mosquito Forceps give you a gentle yet secure grip on delicate tissues. The jaws are serrated for a firm grasp with minimal tissue trauma.
These forceps are designed to be lightweight, minimizing hand fatigue. Due to their small size, mosquito forceps typically call for a fingertip grip technique, where the instrument is held between the fingertips rather than the traditional palm grip. This technique allows for increased sensitivity and control over the forceps' movements. These fine surgical instruments are manufactured from surgical grade stainless steel or premium German steel. They are autoclavable or suitable for chemical sterilization. To preserve the useable life of your surgical instruments, always clean them properly when you finish using them.
Specifications
  • Matière:
    Stainless steel
  • Tip Style:
    Right angle