Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mini Dissecting Scissors, Straight, 9.5 cm | Avantor

Mini Dissecting Scissors, Straight, 9.5 cm

Mini dissecting scissors, fine, sharp round tips, Surgical grade stainless steel, Straight, Large ring handles for people with larger fingers
Catalog # 77663-188
Fournisseur #:  503245
Mini Dissecting Scissors, Straight, 9.5 cm
Catalog # 77663-188
Mini Dissecting Scissors, Straight, 9.5 cm

Mini dissecting scissors, fine, sharp round tips, Surgical grade stainless steel, Straight, Large ring handles for people with larger fingers

Price
$133.83
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Our mini dissecting scissors with Large Rings are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These dissecting scissors have large rings for people with larger fingers.

  • These scissors are 9.5 cm (3.75") long
  • Straight blades with rounded tips and fine blades

These dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. (Standard and medium blades are also available on our mini dissecting scissors).

Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel.

These dissecting scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.

Spécifications
  • Désignation:
    Mini dissecting scissors, fine, sharp round tips
  • Matière:
    Surgical grade stainless steel
  • Curvature:
    Straight
  • Handle:
    Large ring handles for people with larger fingers

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.