Swiss Microdissection Scissors, 9 cm | Avantor
Swiss Microdissection Scissors, 9 cm
Curved, heavy duty, rounded tips, Stainless steel, Sterile
Catalog # 77663-660
Fournisseur #:  504521
Swiss Microdissection Scissors, 9 cm
Catalog # 77663-660
Swiss Microdissection Scissors, 9 cm
Curved, heavy duty, rounded tips, Stainless steel, Sterile
Price
$146.01
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Swiss microdissecting scissors are ring handled surgical scissors particularly suited for dissection and surgical tasks that require precision cutting. These scissors are crafted with high-quality materials and are known for their durability and sharpness. Swiss-made surgical instruments are renowned for their precision and craftsmanship.
  • High quality Swiss-made scissors
  • Sharp, blunt or rounded tips
  • Curved or straight tips
  • Extra fine, fine, or heavy-duty blades
  • Unique beveled design available
  • Swiss surgical grade stainless steel
Swiss manufacturers adhere to strict quality control standards, and their products are known for precision and reliability. WPI Swiss dissecting scissors are made from high-grade stainless steel, which provides corrosion resistance and ensures a long lifespan for the instrument. All these scissors are 9 cm (3.5") long. These scissors have a fine, slender design with sharp blades, allowing for precise cutting in delicate procedures.
They have sharp, blunt or rounded tips These are available with curved or straight tips. You can also get the unique beveled tip design. The handles of Swiss dissecting scissors are designed for ergonomic comfort, providing a secure grip for the surgeon. This is crucial for maintaining control and accuracy during delicate surgical procedures. Like all surgical instruments, Swiss dissecting scissors are designed to be sterilized easily using a chemical sterilization process or by autoclaving.
Spécifications
  • Désignation:
    Curved, heavy duty, rounded tips
  • Matière:
    Stainless steel
  • Sterility:
    Sterile