Swiss Microdissection Scissors, 9 cm | Avantor
Swiss Microdissection Scissors, 9 cm
Curved, extra fine tips, Stainless steel, Sterile
Catalog # 77663-656
Fournisseur #:  504519
Swiss Microdissection Scissors, 9 cm
Catalog # 77663-656
Swiss Microdissection Scissors, 9 cm
Curved, extra fine tips, Stainless steel, Sterile
Price
$138.69
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Swiss microdissecting scissors are ring handled surgical scissors particularly suited for dissection and surgical tasks that require precision cutting. These scissors are crafted with high-quality materials and are known for their durability and sharpness. Swiss-made surgical instruments are renowned for their precision and craftsmanship.
  • High quality Swiss-made scissors
  • Sharp, blunt or rounded tips
  • Curved or straight tips
  • Extra fine, fine, or heavy-duty blades
  • Unique beveled design available
  • Swiss surgical grade stainless steel
Swiss manufacturers adhere to strict quality control standards, and their products are known for precision and reliability. WPI Swiss dissecting scissors are made from high-grade stainless steel, which provides corrosion resistance and ensures a long lifespan for the instrument. All these scissors are 9 cm (3.5") long. These scissors have a fine, slender design with sharp blades, allowing for precise cutting in delicate procedures.
They have sharp, blunt or rounded tips These are available with curved or straight tips. You can also get the unique beveled tip design. The handles of Swiss dissecting scissors are designed for ergonomic comfort, providing a secure grip for the surgeon. This is crucial for maintaining control and accuracy during delicate surgical procedures. Like all surgical instruments, Swiss dissecting scissors are designed to be sterilized easily using a chemical sterilization process or by autoclaving.
Spécifications
  • Désignation:
    Curved, extra fine tips
  • Matière:
    Stainless steel
  • Sterility:
    Sterile
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.