Tenotomy Scissors
Product Details & Documents
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces.
- One serrated blade, one honed razor sharp
- Sharp tips
- Surgical grade stainless steel
These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries.
These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.
Spécifications
- Désignation:Supercut scissor, curved
- Longueur:16.0 cm
- Matière:Stainless steel