Filling Solutions
Filling solution, 10 ml, Oxygen
Catalog # 77663-934
 :  7326
Filling Solutions
Catalog # 77663-934
CAS Number:  
Filling Solutions
Filling solution, 10 ml, Oxygen
CAS Number:  
Price
$158.17
Log in pour la disponibilité
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Filling solutions to use with ISO-NOP, ISO-OXY-2 or OXELP oxygen sensors.
Spécifications
Spécifications
  • Désignation:
    Filling solution
  • Taille:
    10 ml
  • Fill Solution:
    Oxygen