Filling Solutions
Filling Solutions
Filling solution, 10 ml, Nitric oxide
Catalog # 77663-932
Supplier #:  7325
Filling Solutions
Catalog # 77663-932
CAS Number:  
Filling Solutions
Filling solution, 10 ml, Nitric oxide
CAS Number:  
Price
$160.61
Log in pour la disponibilité
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Filling solutions to use with ISO-NOP, ISO-OXY-2 or OXELP oxygen sensors.
Spécifications
Spécifications
  • Désignation:
    Filling solution
  • Taille:
    10 ml
  • Fill Solution:
    Nitric oxide