Noyes Micro Scissors, 12 cm
Noyes micro scissors, blunt/blunt tips, Surgical grade stainless steel, Straight
Catalog # 77663-254
 :  503306
Noyes Micro Scissors, 12 cm
Catalog # 77663-254
Noyes Micro Scissors, 12 cm
Noyes micro scissors, blunt/blunt tips, Surgical grade stainless steel, Straight
Price
$221.42
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions.
  • Spring handle
  • Fine tips for precision surgical applications
These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons.
These scissors are 12 cm (4.7") long and come with sharp/sharp, sharp/blunt, or blunt/blunt tips. On a sharp/sharp pair of scissors, both blades have sharp, pointed tips. These are used for precise cutting of delicate tissues. Use sharp/sharp tips when you need clean, accurate cuts. Blunt/blunt scissors have two blunt tips.
Use these scissors when you need to manipulate tissue during a procedure or dissection and you don't want to risk puncturing or cutting surrounding tissues or structures. A sharp/blunt pair of surgical scissors has one sharp tip and one blunt tip. Use these when you are cutting and manipulating tissues. The sharp tip gives you a precision cut, and you can use the blunt tip for spreading or moving tissues.
Noyes Micro Scissors are made from surgical grade stainless steel or premium German steel, both of which are suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.
Spécifications
  • Désignation:
    Noyes micro scissors, blunt/blunt tips
  • Matière:
    Surgical grade stainless steel
  • Curvature:
    Straight