Microelectrode Holders
Microelectrode holder, size: 2.0 mm
  77664-844
Microelectrode Holders
  77664-844
Microelectrode Holders
Microelectrode holder, size: 2.0 mm
$312.82
Each
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Male connector
  • Pellet half-cell
  • No pressure port
  • Screw cap
Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. The holder style selected will depend on the experimental application, space, and instrumentation.
 : All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Désignation:
    Microelectrode holder, size: 2.0 mm